To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:CD45RO
- Clonality:Monoclonal
- Clone:UCHL1
- Flow Cytometry:Yes
- Isotype:IgG2a
- Reactivity:Primates,Human
- Size:120 tests
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Cultured human T cell line-CA-1.
- Cat. No.:102512-360
- Supplier no.:ANC194050
Human CD45 is expressed on all cells of hematopoietic origin, except erythrocytes. CD45 is a transmembrane tyrosine phosphatase which can exist in at least nine different isoforms resulting from tissue-specific alternative RNA splicing of exons 4-7 of a single gene coding for the various N-terminal peptide segments. The CD45RO isoform contains none of the alternatively spliced exons and is present on most thymocytes, a subpopulation of resting (memory) T cells within both CD4 and CD8 subsets as well as on monocytes and granulocytes. CD45RO plays an essential role in lymphocyte activation.
Type: Primary
Antigen: CD45RO
Clonality: Monoclonal
Clone: UCHL1
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG2a
Reactivity: Human, Primate