To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:MHC2
- Clonality:Monoclonal
- Clone:TDR31.1
- Conjugation:Biotin
- Flow Cytometry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Purified class II from JY cell line.
- Cat. No.:102511-778
- Supplier no.:ANC131030
Specifications
About this item
Human MHC Class II (HLA-DP, DQ & DR) molecules are expressed on dendritic cells, monocytes, macrophages and myeloid beta cells. MHC Class II molecules are heterodimers of polymorphic transmembrane a and b chains and generally bind exogenously derived peptides of about 10-17 amino acids. MHC Class II molecules interact with the T cell receptor on CD4+ T cells.
Type: Primary
Antigen: MHC2
Clonality: Monoclonal
Clone: TDR31.1
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human