To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:TCR
- Clonality:Monoclonal
- Clone:Jovi-1
- Flow Cytometry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Size:120 tests
- Form:liquid
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Human HA1.7 TCR beta chain expressed on transgenic mouse cells.
- Cat. No.:102511-734
- Supplier no.:ANC101040
Specifications
About this item
Human T cell receptor (TCR) is a T cell surface expressed heterodimer of alpha/beta or gamma/delta chains. TCR’s recognize antigen peptides bound to MHC molecules forming the basis of antigen specific response by T cells.
Type: Primary
Antigen: TCR
Clonality: Monoclonal
Clone: Jovi-1
Conjugation: FITC
Epitope:
Host:
Isotype: Mouse IgG2a
Reactivity: Human