Specifications
- Antibody Type:Primary
- Antigen Symbol:MPC-2
- Clonality:Monoclonal
- Clone:JCM-1
- Conjugation:Unconjugated
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:liquid
- Antigen Synonyms:Mitochondrial Pyruvate Carrier 2|Brain Protein 44
- Storage Temperature:–20 °C
- Concentration:1mg/ml
- Shipping Temperature:-20° C
- Immunogen:Peptide coupled to KLH.
- Cat. No.:102971-632
- Supplier no.:AG20B0071C100
Specifications
About this item
Pyruvate is the end-product of glycolysis, a major substrate for oxidative metabolism, and a branching point for glucose, lactate, fatty acid and amino acid synthesis. The mitochondrial enzymes that metabolize pyruvate are physically separated from cytosolic pyruvate pools and rely on a membrane transport system to shuttle pyruvate across the impermeable inner mitochondrial membrane (IMM). Two proteins, mitochondrial pyruvate carriers MPC-1 and MPC-2, form a heterooligomeric complex in the IMM to facilitate pyruvate transport. This step is required for mitochondrial pyruvate oxidation and carboxylation-critical reactions in intermediary metabolism that are dysregulated in several common metabolic diseases.
Type: Primary
Antigen: MPC-2
Clonality: Monoclonal
Clone: JCM-1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG2bkappa
Reactivity: Human, Mouse