Human Recombinant Tartrate-Resistant Acid Phosphatase type 5 (from Baculovirus (Insect cells))
: Avantor
ACP5, also known as tartrate-resistant acid phosphatase type 5, is involved in osteopontin and bone sialoprotein dephosphorylation. Its expression seems to increase in certain pathological states such as Gaucher and Hodgkin diseases, the hairy cell, the B-cell, and the T-cell leukemias. Recombinant human ACP5, fused to His-tag at C-terminus, was expressed in insect cell and purified by using conventional chromatography techniques.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Human
- Storage Conditions:–20 °C
- Biological Activity:Specific activity is > 5,000 units/mg, and is defined as the amount of enzyme that hydrolyze 1.0 nmoles of p-nitrophenyl phosphate (pNPP) per minute at pH 5.0 at 37C.
- Gene ID:54
- Protein Synonyms:SPENCDI|TRAP|Tartrate-resistant acid phosphatase type 5
- Protein/Peptide Name:Tartrate-resistant acid phosphatase type 5
- Molecular Weight:35.1 kDa
- Formulation:Liquid
- Shipping Temperature:Gel Pack
Frequently Bought Together