Anti-ZBTB24 Rabbit Polyclonal Antibody
76011-294
: Prosci
- Antibody Type:Primary
- Antigen Name:Zinc finger and BTB domain-containing protein 24
- Antigen Symbol:ZBTB24
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 ul
- Epitope:Amino Acid Range = 137-166
- Form:Liquid
- Gene ID:O43167
- Antigen Synonyms:ZBTB24
- Modification:None
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Molecular Weight:78 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.4 mg/mL
- Shipping Temperature:4 °C
- Immunogen:This ZBT24 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 137-166 amino acids from the N-terminal region of human ZBT24.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76011-294
ZBTB24 may be involved in BMP2-induced transcription (By similarity).
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:10~50
Type: Primary
Antigen: ZBTB24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Mouse