Anti-FAM155A Rabbit Polyclonal Antibody
76008-962
: Prosci
- Antibody Type:Primary
- Antigen Name:Transmembrane protein FAM155A
- Antigen Symbol:FAM155A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 ul
- Epitope:Amino Acid Range = 45-72
- Form:Liquid
- Gene ID:B1AL88
- Antigen Synonyms:FAM155A
- Modification:None
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Molecular Weight:51 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/mL
- Shipping Temperature:4 °C
- Immunogen:This FAM155A antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 45-72 amino acids from the N-terminal region of human FAM155A.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76008-962
For WB starting dilution is: 1:1000 For FACS starting dilution is: 1:10~50
Type: Primary
Antigen: FAM155A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human