Orders LinkContactUsLinkComponent
GlabrataQuick Kit, Hardy Diagnostics
GlabrataQuick Kit, Hardy Diagnostics
GlabrataQuick Kit, 24
Catalog # 75993-498
Supplier:   Hardy Diagnostics
Supplier #:  Z298
GlabrataQuick Kit, Hardy Diagnostics
Catalog # 75993-498
Supplier:  Hardy Diagnostics
CAS Number:  
GlabrataQuick Kit, Hardy Diagnostics
GlabrataQuick Kit, Hardy Diagnostics
GlabrataQuick Kit, 24
CAS Number:  
Price
$591.00
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
GlabrataQuick™, for the identification of Candida glabrata, 24 tests
  • Test within one to two hours
  • Greater accuracy
  • Easy color read-out
GlabrataQuick™, contains two 96 well trays and 24 Inoculation Buffer tubes with 1ml fill to complete a total of 24 tests. GlabrataQuick™ will accurately identify Candida glabrata within one to two hours. It tests for three carbohydrates for greater accuracy; trehalose, sucrose, and maltose. Easy and distinct color read-out. Carbohydrates turn from blue to yellow.
Specifications
Specifications
  • Description:
    GlabrataQuick Kit
  • No. of tests:
    24