Order Entry
United States
Orders LinkContactUsLinkComponent
 

Nitrate Substrate Broth, Hardy Diagnostics

Nitrate Substrate Broth, Hardy Diagnostics
Supplier:  Hardy Diagnostics
Certificates
Nitrate Substrate Broth, Hardy Diagnostics
Nitrate Substrate Broth, Hardy Diagnostics

3 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

For the determination of nitrate reactions of mycobacteria. Use with AFB Nitrate Reagent A and B

Reagent A contains sulfanilimide, 0.2%.

Reagent A contains sulfanilimide, 0.2%
Reagent B contains naphthylethylenediamine 2HCL, 0.1%

Reagent B contains naphthylethylenediamine 2HCL, 0.1%.

Product Family Options

Product Information

  • Fill volume
    Minimum Order Quantity
    Size
    Pack type
    Availability
    Price
  • 75993-554
    Nitrate Substrate Broth, 13×100 mm
    Nitrate Substrate Broth, 13×100 mm
    2 ml
    5 packs
    13 x 100 mm
    PC tube, optically clear, shatter resistant
    Log in for availability
    Log in for availability
    Pack of 20 (2ml)
    Pack of 20 (2ml) - $131.00
    $131.00
    Pack of 20 (2ml)
       $131.00

    Supplier Number: R97
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 75993-558
    AFB Nitrate Reagent A
    AFB Nitrate Reagent A
    15 ml
    Log in for availability
    Log in for availability
    Each (15ml)
    Each (15ml) - $36.00
    $36.00
    Each (15ml)
       $36.00

    Supplier Number: Z124
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 75993-560
    AFB Nitrate Reagent B
    AFB Nitrate Reagent B
    15 ml
    Log in for availability
    Log in for availability
    Each (15ml)
    Each (15ml) - $30.60
    $30.60
    Each (15ml)
       $30.60

    Supplier Number: Z125
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.