MUG Disks, Hardy Diagnostics
Catalog # 75993-522
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Jar
- Description:MUG Disks
- Size:25 disks/pack
- Cat. No.:75993-522
Specifications
About this item
MUG Disks are recommended for the identification of glucuronidase activity in E.coli by fluorescence, under a long-wave (365nm) UV light source.