Order Entry
United States
ContactUsLinkComponent
SOC Broth, Teknova
SOC Broth, Teknova
Catalog # 100219-988
Supplier:  Teknova
CAS Number:  
SOC Broth, Teknova
Catalog # 100219-988
Supplier:  Teknova
Supplier Number:  S0225
CAS Number:  

Specifications

  • Description:
    SOC Broth
  • Size:
    227 g
  • Cat. No.:
    100219-988
  • Supplier no.:
    S0225

Specifications

About this item

SOC is a nutritionally rich medium used to grow recombinant E.coli

  • Dissolve 32.6 grams per liter of water to reconsitute
  • Autoclave to sterilize
  • Ambient shipping conditions
  • Store at room temperature

Ingredients: 2.0% Tryptone (20 g/L), 0.5% Yeast extract (5.0 g/L), 10 mM Sodium Chloride (NaCl), 2.5 mM Potassium Chloride (KCl), 10 mM Magnesium Chloride (MgCl2), 10 mM Magnesium Sulfate (MgSO4) and 20 mM (0.36%) Glucose (Dextrose) (3.6 g/L).

SOC is the same as SOB, with the addition of Glucose.