ContactUsLinkComponent
Brady® Replacement Ink Pad for BradyJet J5000 Color Label Printer, Brady Worldwide
Brady® Replacement Ink Pad for BradyJet J5000 Color Label Printer, Brady Worldwide
Catalog # 75846-022
Supplier:  Brady Worldwide
undefined
Brady® Replacement Ink Pad for BradyJet J5000 Color Label Printer, Brady Worldwide
Catalog # 75846-022
Supplier:  Brady Worldwide
Supplier Number:  148769
Specifications
  • Description:
    Replacement Ink Pad
  • Dimensions:
    3W×8D×6"H
  • Environmentally Preferable:
  • Cat. No.:
    75846-022
  • Supplier no.:
    J50-PAD
Specifications
About this item
The BradyJet J5000 replacement ink pad keeps the printer in top working condition by absorbing ink waste.
  • Absorbs waste ink to keep the printer clean
  • Waste ink is collected during initial setup and during cleanings
  • Replacement pad is used when initial pad has reached absorption capacity