ContactUsLinkComponent
Brady® Label Roll Adaptor for BMP®71 Label Printer, Brady Worldwide
Brady® Label Roll Adaptor for BMP®71 Label Printer, Brady Worldwide
Catalog # 75840-084
Supplier:  Brady Worldwide
undefined
Brady® Label Roll Adaptor for BMP®71 Label Printer, Brady Worldwide
Catalog # 75840-084
Supplier:  Brady Worldwide
Supplier Number:  147998
Specifications
  • Color:
    Black
  • Description:
    Label roll adaptor for BMP®71 label printer
  • Cat. No.:
    75840-084
  • W×D×H:
    12.7×1.3×8.9 cm
  • Supplier no.:
    M71-ADAPT-BK
Specifications
About this item
Label Roll Adaptors enable the BMP®71 Label Printer to print on BMP®61, TLS 2200®, and HandiMark® Series printer labels and tapes.
  • Package contains three label roll adaptors (BMP®71 Label Printer and label rolls sold separately)
  • Expands the label offering for the BMP®71 Label Printer by more than 600 parts
  • Adaptor snaps on to BMP®61, TLS 2200®, and HandiMark® Series label rolls
This reusable spare adaptors save time by allowing users to keep them on the most frequently used label rolls