ContactUsLinkComponent
Wire Marker Card, Brady
Wire Marker Card, Brady
Catalog # 490006-396
Supplier:  Brady Worldwide
undefined
Wire Marker Card, Brady
Catalog # 490006-396
Supplier:  Brady Worldwide
Supplier Number:  114052
Specifications
  • Color:
    Black
  • Description:
    Wire marker card 1.5"
  • Material:
    Vinyl coated fabric
  • Cat. No.:
    490006-396
  • Supplier no.:
    WM-COL-BK-SC-PK
Specifications
About this item
Repositionable vinyl cloth wire markers from Brady® are perfect for general wire marking needs.
  • Card-mounted wire markers allow for quick, on-the-spot wire marking
  • Excellent flexibility for wrapping around curved surfaces
  • Withstands moderate heat, oil, water, and dirt
Holds tightly to surfaces but allows item to be repositioned with ease if necessary.
Certifications: ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155.