Specifications
- Concentration:1 ml
- Description:Ready-to-Assay™ 5-HT2C Serotonin Family Receptor Frozen Cells
- Size:200 units
- Environmentally Preferable:
- Cat. No.:103382-988
- Host:Chem-1
Specifications
About this item
Human 5-HT2C GPCR frozen cells for Calcium Flux FLIPR Assays.
- Calcium Flux Assay
- Host Chem-1
- Adherent rat hematopoietic cell line
- Expresses endogenous Gα15 protein
5-Hydroxytryptamine (5-HT, also commonly known as serotonin) is synthesized in enterochromaffin cells in the intestine and in serotonergic nerve terminals. In the periphery, 5-HT mediates gastrointestinal motility, platelet aggregation, and contraction of blood vessels. Many functions of the central nervous system are influenced by 5-HT, including sleep, motor activity, sensory perception, arousal and appetite. A family of 12 GPCRs and one ion channel mediate the biological effects of 5-HT (Hoyer et al., 1994). 5-HT2C, which couples to Gq in most cells to stimulate intracellular calcium, is prominently expressed in brain and appears to modulate depression, anxiety and appetite (Miller, 2005; Serretti et al., 2004; Wood, 2003). The mRNA encoding 5-HT2C undergoes selective RNA editing that changes 4 amino acids in the second intracellular loop; these changes result in alteration of efficiency of coupling to G proteins. Alterations in editing of 5-HT2C have been detected in victims of suicidal depression and in mice treated with the SSRI, fluoxetine (Tohda et al., 2006). Cloned human 5-HT2C -expressing cell line is made in the Chem-1 host, which supports high levels of recombinant 5-HT2C expression on the cell surface for functional detection via the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists and modulators at 5-HT2C.