Order Entry
United States
ContactUsLinkComponent
Potassium hydroxide 1% in aqueous solution
Potassium hydroxide 1% in aqueous solution
Catalog # 101410-770
Minority or Woman-Owned Business Enterprise
CAS Number:  
Potassium hydroxide 1% in aqueous solution
Catalog # 101410-770
Supplier Number:  20156-1
Minority or Woman-Owned Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    500 ml
  • Cat. No.:
    101410-770
  • Supplier no.:
    20156-1

Specifications

About this item

 :
 :56.11 g/mol
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553