Ethanol denatured SDA 190 Proof
Product Details & Documents
Decon’s SDA 23A Ethanol, 190 proof is a specially denatured alcohol formulation used for research, healthcare, industrial, or production applications.
While specially denatured alcohol is exempt from Federal Excise Tax, please note that like pure ethanol, Decon’s SDA 23A ethanol, 190 Proof is regulated by the US ATF/TTB. To purchase you must have an Industrial Alcohol User Permit from the AFT/TTB. To apply for this permit visit www.ttb.gov and search for “Form 5150.22” - which is the application for the Industrial Alcohol User Permit.
This product offers high purity and reliability for various laboratory and manufacturing needs, providing consistent performance for cleaning, sanitization, and chemical processing while meeting stringent industry standards.
Packaging: Case (4 X 1 Gallon )
- CAS Number:64-17-5
- Formula:H₃CCH₂OH
- MDL Number:MFCD00003568
- Molecular Weight:46.07 g/mol
Documents
Specifications
Specifications
- Pack type:Plastic bottle
- Size:4×1 gal.
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



