Orders LinkContactUsLinkComponent
Extractor™ Ethidium Bromide Waste Reduction System, GVS Life Sciences
Extractor™ Ethidium Bromide Waste Reduction System, GVS Life Sciences
Extractor™ Ethidium Bromide Waste Reduction System, GVS Life Sciences
Certificates
Extractor™ Ethidium Bromide Waste Reduction System, GVS Life Sciences
2 Options Available Below
Product Details & Documents
One-step filtration polypropylene funnel device for the rapid removal of ethidium bromide from gel-staining solutions.
It contains an activated carbon matrix, which removes >99% of ethidium bromide from electrophoretic buffer quickly and easily
The extractor funnel device fits most standard laboratory flasks and bottles (neck size 33 to 45 mm), and the unit includes a cap for storage between uses. The polypropylene housing is chemically resistant to organics. Pack includes glass fiber prefilters, which remove gel pieces and other debris to avoid premature clogging of the carbon filter.
Each device can decontaminate up to 10 litres of gel-staining solution. The decontaminated solution can be safely poured down the laboratory drain.
Product Family Options
Product Information
  • Availability
    Price
  • 10057-756
    Ethidium Bromide Waste Reduction System
    Ethidium Bromide Waste Reduction System
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $138.03
    $138.03
    Pack of 2
       $138.03
    Supplier Number: 10448030
  • 10057-758
    Ethidium Bromide Waste Reduction System
    Ethidium Bromide Waste Reduction System
    Log in for availability
    Log in for availability
    Pack of 6
    Pack of 6 - $393.74
    $393.74
    Pack of 6
       $393.74
    Supplier Number: 10448031