About this item
Simplify LAMP experiments with these easy-to-use, sensitive and thermostable DNA LAMP master mixes.
- Isothermal Amplification: Facilitates running ampilfication reactions in difficult test environments, enabling use of low tech instrumentation
- Master Mix Format: Simplifies LAMP reaction set up and minimizes optimization required to produce the best assay results
- High Thermal Stability: Highly thermotolerant enzyme (up to 90°C for five minutes) enables pre-heating of reactions containing purified target DNA which, depending on the target, may improve assay sensitivity
- Elevated LAMP Reaction Temperature (68-74°C): Improves primer specificity and reduces background amplification depending on the DNA target
- Freeze-dry Compatible: Enables generation of room temperature stable test kits through lyophilization
The LavaLAMP™ DNA Master Mixes are intended to simplify development and optimization of DNA LAMP (loop-mediated isothermal amplification) reactions. LAMP kits are commonly available as multi-component kits that require optimization (e.g. MgSO4, betaine, enzyme as well as temperature, primer concentration, etc.). The LavaLAMP™ DNA Master Mixes greatly simplify reaction optimization by limiting optimization to target specific components/conditions such as LAMP primer design and reaction temperature. Additionally, these master mixes are heat stable at 90°C for ≤5 minutes, which enables the addition of a reaction pre-heating step which may, depending on the target, increase assay sensitivity and decrease time to results.
It is the sole responsibility of the buyer to ensure that use of the product does not infringe the patent rights of third parties. If the purchaser is not willing to accept these use limitations, Lucigen Corporation is willing to accept return of the product for a full refund.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Environmentally Preferable: