DECON-QUAT® 200V Quaternary Ammonium Solution, Veltek Associates
Supplier: Veltek Associates
Certificates
About this item
Dec-Quat 200V is a neutral pH quaternary ammonium solution It is phosphate free, non-sterile and an EPA registered disinfectant, virucide, sanitizer, germicidal detergent, deodorant and fungicide designed specifically for the life science and research and development industries.
- Clear, Colorless to Straw-Colored Liquid
- 1.01 Specific Gravity
- 6.0 to 8.0 pH @ 1%
- Moderate Foaming
- Excellent Solubility
- Excellent Rinsing
- >100 Viscosity 13.6 mm 2/s
Frequently Bought Together