About this item
Tergitol™ 15-S-7, is a non-ionic Secondary Alcohol Ethoxylate surfactant and is an excellent emulsifier & detergent, enhances paint thickener efficiency and aids in rinseability of solvent-based systems. This product is suggested for use in cleaners, pre-wash spotters and paints & coatings.
- Superior wetting & rinseability
- Excellent formulation & handling properties
- Low odor
- Narrow gel range
- Soluble in water
Density at 20°C (68°F): 0.991 g/ml
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.