About this item
High quality gold nanoparticles. Displaceable citrate surface. Provided with full COA for great value.
- High quality nanoparticles, precisely engineered and highly characterized
- Unagglomerated and monodisperse
- Standard colloidal concentration (0.05 mg/mL, 50 ppm, 0.254 mM, near 1 OD [0.8–1.6 depending on size])
- Citrate surface is stable and displaceable
Spherical gold nanoparticles, 0.05 mg/mL
Each batch is extensively characterized to ensure it meets the highest quality standards, providing reliable and reproducible results. Products are supplied with COA's that include batch-specific TEM, DLS, Zeta Potential, UV-Vis, and pH measurements.
Colloidal gold has applications in Diagnostics, Lateral Flow Assays, Molecular Detection, Photothermal and Microwave-based Therapies, TEM labels, Nanotoxicology, Plasmonics (SPR/Surface Plasmon Resonance), and novel Color Enhancements and Pigments.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.