Order Entry
United States
ContactUsLinkComponent
Synthetic (R)-FSL-1
Synthetic (R)-FSL-1
Catalog # 102981-166
Supplier:  Adipogen
CAS Number:  
Synthetic (R)-FSL-1
Catalog # 102981-166
Supplier:  Adipogen
Supplier Number:  AG-CP3-0010-C100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Vial
  • Protein/Peptide Type:
    Synthetic
  • Size:
    100 μg
  • Storage Conditions:
    Freezer
  • Protein/Peptide Name:
    (R)-FSL-1
  • Purity:
    Absence of detectable protein or DNA contaminants with agonistic TLR activity.
  • Solubility:
    Soluble in water (10mg/ml). Use a homogeniser or sonicator to prepare a homogenous solution.
  • Formulation:
    Colorless powder. Lyophilized
  • Cat. No.:
    102981-166
  • Supplier no.:
    AGCP30010C100

Specifications

About this item

R-FSL-1 (R-Pam2CGDPKHPKSF) is a synthetic lipoprotein that represents the N-terminal part of the 44kDa lipoprotein LP44 of Mycoplasma salivarium. The naturally occurring R-stereoisomer is biologically more active than the S-stereoisomer. Stimulator of TLR2/TLR6. Inducer of TNF-alpha production in macrophages. Upregulates proinflammatory cytokines. Activator of the proinflammatory transcription factor NF-kappaB.

Synonyms:
Fibroblast-stimulating Lipopeptide, R-Pam2CGDPKHPKSF, (R)-Pam2Cys-Gly-Asp-Pro-Lys-His-Pro-Lys-Ser-Phe-OH