About this item
Potent immunosuppressant (same as FK-506 and rapamycin)
Source/Host: Isolated from Tolypocladium inflatum.
Purity: >98%
Appearance/Color: White solid.
Solubility: Soluble in DMSO, ethanol, methanol, dichlormethane, chloroform or acetone. Insoluble in water.
Forms a complex with cyclophilin. Inhibits the activity of the calcium/calmodulin-dependent protein phosphatase 2B (PP2B; calcineurin). Prevents the dephosphorylation of nuclear factor of activated T cells (NFAT) transcription factor, leading to disruption of T cell activation. Suppresses proliferation of cytotoxic T cells and inhibits the production of T cell-derived mediators such as interleukin-2 (IL-2). Prevents rejection of transplanted organs. Anti-inflammatory compound in the treatment of several inflammatory skin diseases (e.g. atopic dermatitis) and with potential anti-rheumatic activity (rheumatoid arthritis). Antibacterial. Antifungal. Antiparasitic. Apoptosis inhibitor. Inhibits the mitochondrial permeability transition pore (MPTP) from opening, thus inhibiting cytochrome c release. NF-kappaB suppressor by induction of unfolded protein response (UPR). Anti-cancer compound. Apoptosis and autophagy inducer. Inhibits nitric oxide synthesis induced by interleukin-1alpha, lipopolysaccharides and TNF-alpha. Potently induces highly cardiogenic progenitors from embryonic stem (ES) cells.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Source:
- Species:
- Environmentally Preferable: