Order Entry
United States
Orders LinkContactUsLinkComponent
SD-06 ≥95%

SD-06 ≥95%

Marketsource
 :   Adipogen
SD-06 ≥95%
 :  Adipogen
Marketsource
 
SD-06 ≥95%

 

 
   
 

SD-06 is a p38 MAP kinase inhibitor. It inhibits p38alpha with an IC(50) value of 170nM and inhibits LPS-stimulated TNF-release in rats.

 :  1-(4-(5-(4-Chlorophenyl)-4-(pyrimidin-4-yl)-1H-pyrazol-3-yl)piperidin-1-yl)-2-hydroxyethan-1-one, SD 0006
  •  :271576-80-8
  •  :C₂₀H₂₀ClN₅O₂
  •  :MFCD17019341
  •  :397.9 g/mol
  •  :Freezer
 

 

  • Purity:
    95%
  • Appearance/Color:
    Solid.
  • Solubility:
    Soluble in DMSO.
Product Family Options

 

  • Size
    Pack type
     
     
  • 1 mg
    Plastic vial
       
       
    Each (1mg)
    Each (1mg) - $205.00
    $205.00
    Each (1mg)
       $205.00

     : SYN-1078-M001
     
     
    Marketsource
    Marketsource
  • 5 mg
    Plastic vial
       
       
    Each (5mg)
    Each (5mg) - $416.00
    $416.00
    Each (5mg)
       $416.00

     : SYN-1078-M005
     
     
    Marketsource
    Marketsource
  • 10 mg
    Plastic vial
       
       
    Each (10mg)
    Each (10mg) - $653.00
    $653.00
    Each (10mg)
       $653.00

     : SYN-1078-M010
     
     
    Marketsource
    Marketsource

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.