About this item
Mitochondria-derived peptide (MDP). Promotes biosynthesis of the endogenous AMP analog AICAR and consequently AMP-activated protein kinase (AMPK). Induces cellular and systemic glucose uptake and improves insulin sensitivity. Shown to prevent diet-induced obesity (DIO) in mice. Potential anti-obesity and anti-aging compound.
- Solid lyophilized powder
- Soluble in water
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- CAS No.:1627580-64-6
- Source:Synthetic
- Species:Human
- Storage Conditions:Freezer
- Protein/Peptide Name:MOTS-c
- Purity:>95% (HPLC)
- Molecular Weight:2174.6 + 114.0
- Molecular Formula:C₁₀₁H₁₅₂N₂₈O₂₂S₂ · C₂HF₃O₂