clasto-Lactacystin beta-lactone ≥95% (by HPLC)
Catalog # 102988-280
Supplier: Adipogen
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Vial
- Size:100 µg
- Cat. No.:102988-280
- Supplier no.:AGCN20442C100
Specifications
Specification Test Results
- Purity:95% (HPLC)
- Appearance/Color:White to off-white solid.
- Solubility:Soluble in DMSO or acetonitrile. Undergoes hydrolysis in aqueous buffers.
Specification Test Results
About this item
:
:213.2 g/mol
Storage Temperature:Freezer
CAS Number:154226-60-5
Formula:C₁₀H₁₅NO₄
MDL Number:MFCD01076526