
Okadaic acid potassium salt ≥98% (by HPLC), high purity
Catalog # 102987-192
Supplier: Adipogen
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Vial
- Size:100 µg
- Cat. No.:102987-192
- Supplier no.:AGCN20060C100
Specifications
Specification Test Results
- Source/Host:Isolated from Prorocentrum concavum. Salt form generated in aqueous potassium hydroxide-methanol solution.
- Purity:98% (HPLC)
- Appearance/Color:White to off-white solid.
- Solubility:Soluble in DMSO, ethanol, methanol, acetone, ethyl acetate or distilled water. Dissolve first in organic solvent before diluting in water.
Specification Test Results
About this item
:
ADR:6.1,III
:843.11 g/mol
UN:3462
:GHS06
Storage Temperature:Freezer
CAS Number:155751-72-7
:209266-79-5
Formula:C₄₄H₆₇O₁₃·K
MDL Number:MFCD00214356