
Okadaic acid ammonium salt ≥98% (by HPLC), high purity
Supplier: Adipogen
Certificates
About this item
CAS Number:155716-06-6
:175522-42-6
Formula:C₄₄H₆₇O₁₃·NH₄
MDL Number:MFCD00153856
:822.03 g/mol
Storage Temperature:Freezer
UN:3462
ADR:6.1,III
:GHS06
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Source/Host:Isolated from Prorocentrum concavum. Salt form generated in aqueous ammonium hydroxide-methanol solution.
- Purity:98% (HPLC)
- Appearance/Color:White to off-white solid.
- Solubility:Soluble in DMSO, ethanol, methanol, acetone, ethyl acetate or distilled water. Dissolve first in organic solvent before diluting in water.
Specification Test Results
Frequently Bought Together