ContactUsLinkComponent
REPEATER Pump Tube Set, Baxter IV Solutions
REPEATER Pump Tube Set, Baxter IV Solutions
Catalog # 10837-306
Supplier:  Baxter
REPEATER Pump Tube Set, Baxter IV Solutions
Catalog # 10837-306
Supplier:  Baxter
Supplier Number:  H93811
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Fluid Transfer Tube Set with Vented Spike, Clear
  • Length:
    210 mm
  • Cat. No.:
    10837-306
  • Supplier no.:
    H93811
Specifications
About this item
REPEATER pump tube set with universal spike-luer lock.
  • Sterile
  • Rx only
It consists of inlet spike and male luer lock outlet for transfer of sterile fluids from one container to another using the REPEATER Pump.
Delivery: Includes RAPIDFILL Connectors, Dispensing Pins or TWOFER Needles.