ContactUsLinkComponent
Disposable Mop Head, Contec®
Disposable Mop Head, Contec®
Catalog # 47734-202
Supplier:  Contec
Small Business Enterprise
undefined
Disposable Mop Head, Contec®
Catalog # 47734-202
Supplier:  Contec
Supplier Number:  C3-MOP
Small Business Enterprise
Specifications
  • Description:
    Disposable Mop Head
  • Environmentally Preferable:
  • Cat. No.:
    47734-202
  • Supplier no.:
    C3-MOP
Specifications
About this item
Made from 100% polyester fabric, the non-woven structure of hydroentangled fibers traps particulates until removed by a wringer.
  • Provides excellent sorbency
  • Blue band for visibility
  • Sterilizable by autoclave, ethylene oxide, or gamma radiation
  • Can be used with fiberglass handle