International Polymer Solutions PTFE Purge Adapter with Shutoff, Ace Glass Incorporated
: Avantor
PTFE Purge/Shutoff Ace-Thred adapter allows the purging of air-sensitive contents through the use of two top taps that are controlled by a two-way stopcock.
- PTFE construction
- Connects via an Ace-Thred
- Two top taps
- Two-way stopcock
The PTFE construction allows for higher and lower temperature use than most other plastics. Adapter connects via an Ace-Thred, which is an internal thread used in conjunction with an O-Ring. It features two top taps controlled by a two-way stopcock. Max. 160 psig, Min. 0.003 mm Hg.
Frequently Bought Together