Order Entry
United States
Orders LinkContactUsLinkComponent
Calcium Levofolinate USP Reference Standard

Calcium Levofolinate USP Reference Standard

500 mg
Catalog # 101096-464
Marketsource
Supplier:   USP
Supplier #:  1358004
Calcium Levofolinate USP Reference Standard
Catalog # 101096-464
Supplier:  USP
Marketsource
CAS Number:  80433-71-2
Calcium Levofolinate USP Reference Standard

500 mg

CAS Number:  80433-71-2
Price
$475.38
Log in for availability
Each (500mg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  (4-((((S)-2-Amino-5-formyl-4-oxo-1,4,5,6,7,8-hexahydropteridin-6-yl)methyl)amino)benzoyl)-L-glutamic acid, calcium salt(1:1), Levoleucovorin calcium
  • CAS Number:80433-71-2
  • Formula:C₂₀H₂₁CaN₇O₇
  • MDL Number:MFCD08704249
  • Molecular Weight:511.50 g/mol
Specifications

Specifications

  • Size:
    500 mg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.