Order Entry
United States
ContactUsLinkComponent
Diluent for Maximum Recovery, HiMedia Laboratories
Diluent for Maximum Recovery, HiMedia Laboratories
Supplier:  HiMedia
Certificates
Diluent for Maximum Recovery, HiMedia Laboratories
Supplier:  HiMedia
Certificates

Specifications

  • Storage Temperature:
    35-37 °C
  • pH:
    6.80-7.20

Specifications

Related Information

Frequently Bought Together