ContactUsLinkComponent
In-line Filter Traps, PerkinElmer
In-line Filter Traps, PerkinElmer
Catalog # 97013-134
Supplier:  PerkinElmer
undefined
In-line Filter Traps, PerkinElmer
Catalog # 97013-134
Supplier:  PerkinElmer
Supplier Number:  N9301178
Specifications
  • Description:
    In-line Filter trap
  • Material:
    Stainless steel
  • Cat. No.:
    97013-134
  • Supplier no.:
    N9301178
Specifications
About this item
Pre-instrument in-line filter trap for removal of moisture, oil, and dust from GC input gases.