Order Entry
United States
ContactUsLinkComponent
Acid red 52
Acid red 52
Catalog # 700009-970
Small Business Enterprise
CAS Number:  
Acid red 52
Catalog # 700009-970
Supplier Number:  A1586-25GM
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass, amber
  • Size:
    25 g
  • Cat. No.:
    700009-970
  • Supplier no.:
    A1586-25GM

Specifications

About this item

 :
 :580.65 g/mol
Storage Temperature:Ambient
CAS Number:3520-42-1
Formula:C₂₇H₃₀N₂O₇S₂
MDL Number:MFCD00010180