Rheodyne™ 7725 Analytical Injection Valve, PerkinElmer
Rheodyne™ 7725 and 7725i valves are versatile injectors and can use both partial-filling and complete-filling methods for loading the sample loop.
- Improved access to fittings
- Inject from 1μL to 5mL with high accuracy and precision
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.