Silica Reagents for 5000 Silica Analyzer, Hach
Supplier: Avantor
Certificates
About this item
One unit each of the reagents is sufficient for 30 days of operation
The standard solution is sufficient for 10 calibrations.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.