Order Entry
United States
ContactUsLinkComponent
Single Element Standards, Aluminium, for ICP-MS, Pure Plus grade, PerkinElmer
Single Element Standards, Aluminium, for ICP-MS, Pure Plus grade, PerkinElmer
Catalog # 97011-980
Supplier:  PerkinElmer
CAS Number:  
undefined
Single Element Standards, Aluminium, for ICP-MS, Pure Plus grade, PerkinElmer
Catalog # 97011-980
Supplier:  PerkinElmer
Supplier Number:  N9300184
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml Al
  • Matrix:
    2% HNO₃
  • Size:
    125 ml
  • Cat. No.:
    97011-980
  • Element:
    Aluminum
  • Supplier no.:
    N9300184

Specifications

About this item

  • Analyzed by ICP-OES
  • Analyzed by classical wet assay
  • 32 trace impurities analyzed by ICP-MS of the final solution
  • Impurities reported at ppm level
Certifications: ISO 17034 and ISO 17025, certified by A2LA
Delivery: Supplied with a certificate of analysis that documents the quality and reliability.