Mega Tube, Watson Bio Lab
75982-894
- Cap type:Slight Stiff-Touch cap
- Volume:5.0 ml
- Description:Natural Megatube with Round Bottom
- Cat. No.:75982-894
These large-volume centrifuge tubes feature a milliliter scale and a snap cap.
- Flat snap cap
- Multiple marking spaces
- Autoclavable polypropylene construction
- RNase-/DNase-/Human DNA-free
- Large volume
Users can mark the top of the flat snap cap. More marking space is available on the tube itself. The clear, autoclavable polypropylene material doesn't contain DNase, RNase, or human DNA.