PCR Plates, Watson Bio Lab
Supplier: Watson Bio Lab
Jump to:
About this item
The wells are uniformly thin, which enables PCR with superior heat exchange.
- Precast Demarcation Lines
- Thin Wall
- High Transparency
- Nuclease-free Certified
- Autoclavable Polypropylene Construction
These PCR plates feature autoclavable polypropylene construction and precast demarcation lines. Users can cut the plates as needed.
3 Options Available Below
Product Details & Documents
The wells are uniformly thin, which enables PCR with superior heat exchange.
- Precast Demarcation Lines
- Thin Wall
- High Transparency
- Nuclease-free Certified
- Autoclavable Polypropylene Construction
These PCR plates feature autoclavable polypropylene construction and precast demarcation lines. Users can cut the plates as needed.
Product Family Options
Product Information
- No. of wellsSkirtWell volumeAvailabilityPrice
- 96Non-Skirted0,2 mlBox of 50$187.53Case of 250$937.65Supplier Number: 137-174C
- 96Non-Skirted0,1 mlBox of 10$68.65Case of 300$2,059.50Supplier Number: 137-674C
- 96Semi-Skirted0,1 mlBox of 10$68.35Case of 300$2,050.50Supplier Number: 137-675C