Order Entry
United States
ContactUsLinkComponent
Pierce QuantaBlu™ NS/K Fluorogenic Substrate Kit, Thermo Scientific
Pierce QuantaBlu™ NS/K Fluorogenic Substrate Kit, Thermo Scientific
Catalog # PI15162
Supplier:  Invitrogen
CAS Number:  
Pierce QuantaBlu™ NS/K Fluorogenic Substrate Kit, Thermo Scientific
Catalog # PI15162
Supplier:  Invitrogen
Supplier Number:  15162
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Cat. No.:
    PI15162
  • Volume:
    280 ml
  • Supplier no.:
    15162

Specifications

About this item

QuantaBlu™ NS/K fluorogenic substrate kit is a two-component kit containing proprietary fluorescent substrate and peroxide solutions for non-stopped kinetic assays

  • More sensitive than TMB, OPD or ABTS substrates
  • Large dynamic range (4 log peroxidase concentration range)
  • Working solution stable for 24 hours
  • Large stokes shift; excitation/emission maxima of 325/420; range of 315 to 345/370 to 460
  • Does not photobleach

QuantaBlu™ substrate exhibits a flat baseline in assays, which facilitates low-level detection sensitivity and allows for high signal-to-noise ratios. 275 mL of working solution is sufficient for 2750 wells at 100 µL per well.

Delivery: Kit includes QuantaBlu™ substrate solution (250 mL), QuantaBlu™ peroxide solution (30 mL).