Specifications
- Description:Endothermic Reactions Lab Kit
- No. of lab groups:15
- Cat. No.:470225-904
- Supplier no.:AD-WE2500-KIT
Specifications
About this item
Monitor and graph temperature change in an endothermic reaction.
- Hands on lab activity
- Kit aligned to next generation standards MS-PS1-2
- Instructions and materials included for 15 groups
- Middle school
In this lab, students will monitor and graph temperature change in an endothermic reaction, and create a cold pack, which utilizes and endothermic reaction.