ContactUsLinkComponent
Ward's® Essentials Catenary Arch Kit
Ward's® Essentials Catenary Arch Kit
Catalog # 470225-366
Supplier:  Wards
undefined
Ward's® Essentials Catenary Arch Kit
Catalog # 470225-366
Supplier:  Wards
Supplier Number:  VWRGP00001
Specifications
  • Description:
    Catenary Arch Kit
  • No. of lab groups:
    1
  • Cat. No.:
    470225-366
  • Supplier no.:
    VWRGP00001
Specifications
About this item
Students will explore the Catenary Arch and it's forces.
  • Hands on lab activity
  • Kit aligned to next generation standards MS-PS2-4, MS-PS2-5
  • Instructions and materials included for 1 group
  • Middle school
The Standing Cantenary Arch Kit is composed of seven wooden blocks that assemble to produce the shape of the catenary arch. This kit engages students to learn engineering concepts such as balancing force and load transfer through the assembly of the arch.