Orders LinkContactUsLinkComponent
EpiScript™ RNase H- Reverse Transcriptase, cDNA synthesis, Biosearch Technologies
EpiScript™ RNase H- Reverse Transcriptase, cDNA synthesis, Biosearch Technologies
Supplier:   Lucigen
EpiScript™ RNase H- Reverse Transcriptase, cDNA synthesis, Biosearch Technologies
Supplier:  Lucigen
Certificates
EpiScript™ RNase H- Reverse Transcriptase, cDNA synthesis, Biosearch Technologies
EpiScript™ RNase H- Reverse Transcriptase, cDNA synthesis, Biosearch Technologies
2 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
An optimized MMLV reverse transcriptase enzyme for improved production of full-length cDNA from limiting total RNA samples.
  • Efficient and Sensitive: Produce full-length cDNA from as little as 50 pg of total RNA
  • Improved Specificity: Run reaction temperatures as high as 55°C to enhance primer specificity and improve transcription of GC-rich or high secondary structure target RNAs
EpiScript™ RNase H- Reverse Transcriptase (EpiScript RT), an alternative to SuperScript® II Reverse Transcriptase, is a recombinant MMLV reverse transcriptase with greatly reduced RNase H activity. It is highly efficient at producing full-length cDNA from long RNA templates and is capable of producing cDNA from as little as 50 pg of total RNA for real-time RT-PCR (qRT-PCR) analysis and other applications.
Delivery information: Contains EpiScript RNase H- Reverse Transcriptase, 10X Reaction Buffer and 100 mM DTT.
Caution: For research use only. Not for human or diagnostic use.
Documents
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 75927-940
    EpiScript™ RNase H- Reverse Transcriptase
    EpiScript™ RNase H- Reverse Transcriptase
    10000 Units
    Log in for availability
    Log in for availability
    Each
    Each - $268.86
    $268.86
    Each
       $268.86
    Minimum order quantity is 1
    Supplier Number: ERT12910K
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 75927-942
    EpiScript™ RNase H- Reverse Transcriptase
    EpiScript™ RNase H- Reverse Transcriptase
    25000 Units
    Log in for availability
    Log in for availability
    Each
    Each - $450.02
    $450.02
    Each
       $450.02
    Minimum order quantity is 1
    Supplier Number: ERT12925K
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.