ContactUsLinkComponent
Go Direct™ ORP Electrode
Go Direct™ ORP Electrode
  470166-520
undefined
Go Direct™ ORP Electrode
  470166-520
 :  GDX-ORP-BNC
  • Description:
    Go Direct™ ORP electrode
  • Cat. No.:
    470166-520
  • Supplier no.:
    GDX-ORP-BNC
The Go Direct ORP (Oxidation-Reduction Potential) Sensor measures the ability of a solution to act as an oxidizing or reducing agent.
  • The Go Direct ORP Sensor can be used in a variety of experiments
  • Determine the equivalence point of an oxidation-reduction titration
  • Measure the oxidizing ability of chlorine in swimming pools
  • Investigate the ORP of drinking water
Use the Go Direct ORP Sensor to measure the ability of a solution to act as an oxidizing agent and to quantify ion activity. This sensor measures the capacity of a solution to either release or accept electrons from chemical reactions.
This sensor is comprised of an ORP electrode connected to a Go Direct Electrode Amplifier via a BNC connector. This design enables you to use the Go Direct Electrode Amplifier with other Vernier electrodes, such as the pH Electrode, Flat pH Electrode, ORP Electrode, ISE electrode, or a third-party electrode.
The Go Direct robust rechargeable battery offers long battery life and provides always-ready operation. Monitor battery life directly from our free Graphical Analysis™ 4 app. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting—no interruptions or inconvenience to you or your students.
Our Go Direct™ ORP Sensor connects directly to your mobile device, Chromebook, or computer using our free Graphical Analysis™ 4 app—no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth, allowing you to choose the solution best for your classroom or laboratory.