ContactUsLinkComponent
CappWash™ Microplate Washer, APS BE
  490015-894
CappWash™ Microplate Washer, APS BE
  490015-894
 :  W-12KIT-115V
  • Description:
    CappWash™ Kit; Includes W-12, WP-115V, WB-1, WB-3, WB-4, and W-1000 (3 meters)
  • Cat. No.:
    490015-894
  • Supplier no.:
    W-12KIT-115V
CappWash™ is the ideal washing tool for small scale immuno assays
  • Compact and made for small-scale work
  • Unique two-in-one design
  • Simple washing; matches all well types
  • No programming and no electrical wiring connection required
The vacuum pump is available for 115V connections.
Fitting into even the tightest budget, CappWash™ is a perfect alternative for washing micro plates and strips.