Karl Fischer Reagents, Honeywell Research Chemicals
Supplier: Honeywell Research Chemicals
Certificates
About this item
Pyridine base KF reagents.
Mixture of solutions A and B (ratio 1:1) yield water equivalent (titer) of mi, for pyridine-based volumetric KF titration (iodine, methanol)
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.