About this item
SensiCare with aloe surgical gloves are manufactured with isolex polyisoprene, ensuring a safe, high quality alternative to natural rubber latex.
- The anatomically curved style provides comfort
- Anti-roll-down cuff technology significantly reduces the roll down common with many surgical gloves
- Gloves contain freeze-dried aloe
Coated with 100% pure aloe vera to help soothe and moisturize dry, irritated skin caused by frequent hand washing and glove-wearing.
Ideal for general surgery.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 54720-112
- 54720-116
- 54720-114
- 54720-110
- 89068-646
- 89068-644
- 89068-648
- Latex/Latex-free:
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Powdered/Powder-free:
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Finish:
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Cuff Style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Finger Thickness:
- 9.8 mils
- 9.8 mils
- 9.8 mils
- 9.8 mils
- 9.8 mils
- 9.8 mils
- 9.8 mils