


Toluene ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # BJ650579-1L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:1 L
- Cat. No.:BJ650579-1L
- Supplier no.:650579-1L
Specifications
Specification Test Results
- IR spectrum:complying
- Assay (GC):Min. 99.90 %
- GC (as heptachlor epoxide):Max. 10 ng/l
- Non-volatile matter:Max. 0.0001 %
- Water (Karl Fischer):Max. 0.02 %
- Absorbance at 284 nm:Max. 1.0
- Absorbance at 300 nm:Max. 0.10
- Absorbance at 325 nm:Max. 0.02
- Absorbance at 350 nm:Max. 0.005
- Absorbance at 400 nm:Max. 0.005
- Autoignition Temperature:997 °F
- Vapor Pressure:22 mmHg ( 20 °C)
Specification Test Results
About this item
:
:92.14 g/mol
Boiling Pt:110.6 °C
Melting Pt:–95 °C
Density:0.867 g/cm³ (20 °C)
Flash Pt:7.1 %(V)
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
:GHS07
:GHS08
:GHS02
CAS Number:108-88-3
Formula:C₆H₅CH₃
MDL Number:MFCD00008512